Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ US Country Box for the Milli-Q™ IQ 7000 Ultrapure Water System

Catalog No. ZFCBIQ0US Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Milli-Q IQ 7000 Ultrapure Water System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
ZFCBIQ0US Milli-Q IQ 7000 Ultrapure Water System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZFCBIQ0US Supplier MilliporeSigma™ Supplier No. ZFCBIQ0US
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A US specific country box with the user manual in local language and a power cord adapted to the local electrical network

Order Info

Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
Type Accessory to Milli-Q IQ 7000 Ultrapure Water System
Purity Meter Included No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.