Learn More
Innovative Research Inc US Cow Aqes Hmr Fz 100ml
Catalog No. 502744586
Encompass_Preferred
Innovative Grade US Origin Bovine Aqueous Humor Frozen 100ml - Innovative Grade US Origin Bovine Aqueous Humor from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Aqueous Humor refers to the fluid in the eye. This is processed from animals younger than 30 months of age.This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4586 Supplier Innovative Research Inc Supplier No. IGBOAQHZ100ML
Innovative Grade US Origin Bovine Aqueous Humor Frozen 100ml - Innovative Grade US Origin Bovine Aqueous Humor from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Aqueous Humor refers to the fluid in the eye. This is processed from animals younger than 30 months of age.This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).