Learn More
Innovative Research Inc US Cow Articular Crtlg Fz 10gm
Catalog No. 502744588
Encompass_Preferred
Innovative Grade US Origin Bovine Articular Cartilage Frozen 10gm - Innovative Grade US Origin Bovine Articular Cartilage from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided.This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4588 Supplier Innovative Research Inc Supplier No. IGBOACRCZ10GM
Innovative Grade US Origin Bovine Articular Cartilage Frozen 10gm - Innovative Grade US Origin Bovine Articular Cartilage from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided.This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).