Learn More
Innovative Research Inc US Cow Calf Knee Fz
Catalog No. 502744551
Encompass_Preferred
Innovative Grade US Origin Bovine Calf Knee Frozen - Innovative Grade US Origin Bovine Calf Knee from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4551 Supplier Innovative Research Inc Supplier No. IGCLFKNZ
Innovative Grade US Origin Bovine Calf Knee Frozen - Innovative Grade US Origin Bovine Calf Knee from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).