Learn More
Innovative Research Inc US Cow Calf Knee Fz
Supplier:  Innovative Research Inc IGCNBGCSER5ML
Encompass_Preferred
Innovative Grade US Origin Canine Beagle Complement Serum 5ml - Innovative Grade US Origin Canine Beagle Complement Serum from Innovative Research is manufactured using whole blood collections from Beagles. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity - custom volumes are available upon request. This product is ideal for hemolytic assay where hemolysis of IgG antibodies are required. This complement serum is preservative-free.
Catalog No. 50-274-4550