Learn More
Innovative Research Inc US Cow Eye Retina Fz
Catalog No. 502744579
Encompass_Preferred
Innovative Grade US Origin Bovine Eye Retina Frozen - Innovative Grade US Origin Bovine Eye Retina from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. The adult bovine product is processed from animals less than 30 months of age. The calf product is processed from animals less than 12 months of age. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4579 Supplier Innovative Research Inc Supplier No. IGBOEYRZ
Innovative Grade US Origin Bovine Eye Retina Frozen - Innovative Grade US Origin Bovine Eye Retina from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. The adult bovine product is processed from animals less than 30 months of age. The calf product is processed from animals less than 12 months of age. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).