Learn More
Innovative Research Inc US Cow Tendon Extensor Fz
Catalog No. 502744564
Encompass_Preferred
Innovative Grade US Origin Bovine Tendon Extensor Frozen - Innovative Grade US Origin Bovine Tendon Extensor from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Available from adult (fewer than 30 months of age; tendon measures 4 to 6 inches in length), calf (fewer than 12 months of age), and fetal (tendon measures 3 to 4 inches in length) bovine.This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4564 Supplier Innovative Research Inc Supplier No. IGBOTDEXZ
Innovative Grade US Origin Bovine Tendon Extensor Frozen - Innovative Grade US Origin Bovine Tendon Extensor from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Available from adult (fewer than 30 months of age; tendon measures 4 to 6 inches in length), calf (fewer than 12 months of age), and fetal (tendon measures 3 to 4 inches in length) bovine.This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).