Learn More
Innovative Research Inc US Cow Tndn Deep Dgtl Flexr Fz
Catalog No. 502744565
Encompass_Preferred
Innovative Grade US Origin Bovine Tendon Deep Digital Flexor Frozen - Innovative Grade US Origin Bovine Tendon Deep Digital Flexor from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Measures approximately 8 to 10 inches in length (in an adult animal), and is processed to include the two connecting foot tendons. Available from adult (fewer than 30 months of age), calf (fewer than 12 months of age), or fetal bovine. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4565 Supplier Innovative Research Inc Supplier No. IGBOTDDFZ
Innovative Grade US Origin Bovine Tendon Deep Digital Flexor Frozen - Innovative Grade US Origin Bovine Tendon Deep Digital Flexor from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Measures approximately 8 to 10 inches in length (in an adult animal), and is processed to include the two connecting foot tendons. Available from adult (fewer than 30 months of age), calf (fewer than 12 months of age), or fetal bovine. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).