Learn More
Innovative Research Inc US Cow Whl Bld NaCt 100ml
Catalog No. 502744558
Encompass_Preferred
Innovative Grade US Origin Bovine Whole Blood Na Citrate 100ml - Innovative Grade US Origin Bovine Whole Blood from Innovative Research is collected per-order from cows at USDA-approved facilities . The animals used for this material are of US Origin, ensuring that only the highest-quality material is used.
Catalog No. 50-274-4558 Supplier Innovative Research Inc Supplier No. IGBOWBNAC100ML
Innovative Grade US Origin Bovine Whole Blood Na Citrate 100ml - Innovative Grade US Origin Bovine Whole Blood from Innovative Research is collected per-order from cows at USDA-approved facilities . The animals used for this material are of US Origin, ensuring that only the highest-quality material is used.