Learn More
Rocky Mountain Biologicals, LLC RMBIO Heat Inactivated, US Origin, 500ML
SDP
Catalog No. NC1928571
Encompass_Preferred
RMBIO FBS is sourced from USDA-approved suppliers and is processed in our cGMP-compliant and FDA-registered facility. Our FBS is heat inactivated at 56C for 30 minutes. RMBIO FBS is processed under strict protocols that ensure sterility, high quality, and excellent growth-promotion and performance over a range of applications and cell lines. Heat inactivatedLow endotoxin and hemoglobin levelsExcellent growth promotion characteristicsAverage endotoxin level*: < 1 EU/mLAverage hemoglobin level*: < 15 mg/dLTotal protein: 3.0-4.5 g/dLTriple 0.1 m sterile filtration9CFR-tested US originStorage conditions: -10 C * Data based on last 10 lots produced.
Catalog No. NC1928571 Supplier Rocky Mountain Biologicals, LLC Supplier No. FBSBHT5XM
RMBIO FBS is sourced from USDA-approved suppliers and is processed in our cGMP-compliant and FDA-registered facility. Our FBS is heat inactivated at 56C for 30 minutes. RMBIO FBS is processed under strict protocols that ensure sterility, high quality, and excellent growth-promotion and performance over a range of applications and cell lines. Heat inactivatedLow endotoxin and hemoglobin levelsExcellent growth promotion characteristicsAverage endotoxin level*: < 1 EU/mLAverage hemoglobin level*: < 15 mg/dLTotal protein: 3.0-4.5 g/dLTriple 0.1 m sterile filtration9CFR-tested US originStorage conditions: -10 C * Data based on last 10 lots produced.