Learn More
Innovative Research Inc US Horse Bld Defibrin 500ml
Supplier:  Innovative Research Inc IGHSWBDF500ML
Encompass_Preferred
Innovative Grade US Origin Horse Blood Defibrinated 500ml - Innovative Grade US Origin Horse Whole Blood Defibrinated from Innovative Research is collected per-order from horses in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color.
Catalog No. 50-274-4508