Learn More
Bruker Daltonics US IVD BTS(BACTERIAL TEST STND
Catalog No. NC0884024
Encompass_Preferred
US IVD Bacterial Test Standard. Mass calibration standard containing a typical Escherichia coli DH5 alpha peptide and protein profile plus additional proteins. For quality control and validation of MALDI Biotyper CA Systems.
Catalog No. NC0884024 Supplier Bruker Daltonics Supplier No. 8604530
US IVD Bacterial Test Standard. Mass calibration standard containing a typical Escherichia coli DH5 alpha peptide and protein profile plus additional proteins. For quality control and validation of MALDI Biotyper CA Systems.