Learn More
Bruker Daltonics US IVD HCCA PORTIONED
Catalog No. NC0884023
Encompass_Preferred
US IVD HCCA portioned (referred to as 'US IVD HCCA'; HCCA = α-Cyano-4-hydroxycinnamic acid) enables easy and convenient preparation of MALDI matrix solution for MALDI Biotyper CA measurement and identification of test organisms. US IVD HCCA is soluble in standard solvent (see section 4). Each tube contains 2.5±0.3 mg matrix, enabling fast preparation of the desired concentration simply by adding the recommended volume of standard solven
Catalog No. NC0884023 Supplier Bruker Daltonics Supplier No. 8604531
US IVD HCCA portioned (referred to as 'US IVD HCCA'; HCCA = α-Cyano-4-hydroxycinnamic acid) enables easy and convenient preparation of MALDI matrix solution for MALDI Biotyper CA measurement and identification of test organisms. US IVD HCCA is soluble in standard solvent (see section 4). Each tube contains 2.5±0.3 mg matrix, enabling fast preparation of the desired concentration simply by adding the recommended volume of standard solven