Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Power cord assembly US&JP plug 16A 125V

Catalog No. NC1627760 Shop All Thermo Scientific Products
Encompass_Preferred

Power cord assembly US&JP plug 16A 125V

Catalog No. NC1627760 Supplier Thermo Scientific Supplier No. CIC0001471
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Power cord assembly US&JP plug 16A 125V

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.