Learn More
Innovative Research Inc US Mnky Cyno Comp Srm 100ml
Catalog No. 502744502
Encompass_Preferred
Innovative Grade US Origin Monkey Cynomolgus Complement Serum 100ml - Innovative Grade US Origin Monkey Cynomolgus Complement Serum from Innovative Research is manufactured using whole blood collections from type Cynomolgus monkeys. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.
Catalog No. 50-274-4502 Supplier Innovative Research Inc Supplier No. IGMNCYCSER100ML
Innovative Grade US Origin Monkey Cynomolgus Complement Serum 100ml - Innovative Grade US Origin Monkey Cynomolgus Complement Serum from Innovative Research is manufactured using whole blood collections from type Cynomolgus monkeys. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.