Learn More
Innovative Research Inc US Ms C57 Comp Srm 25ml
Catalog No. 502744479
Encompass_Preferred
Innovative Grade US Origin Mouse C57BL6 Complement Serum 25ml - Innovative Grade US Origin Mouse C57BL6 Complement Serum from Innovative Research is manufactured using whole blood collections from type C57BL6 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.
Catalog No. 50-274-4479 Supplier Innovative Research Inc Supplier No. IGMSC57CSER25ML
Innovative Grade US Origin Mouse C57BL6 Complement Serum 25ml - Innovative Grade US Origin Mouse C57BL6 Complement Serum from Innovative Research is manufactured using whole blood collections from type C57BL6 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.