Learn More
Innovative Research Inc US Ms C57 Sngl Whl Bld K2E Vl
Catalog No. 502744464
Encompass_Preferred
Innovative Grade US Origin Mouse C57BL6 Single Donor Whole Blood K2 EDTA Vial - Innovative Grade US Origin Mouse C57BL6 Single Donor Whole Blood from Innovative Research is collected per order from type C57BL6 Mice. The animals used for this material are of US Origin, ensuring the highest-quality material.
Catalog No. 50-274-4464 Supplier Innovative Research Inc Supplier No. IGMSC57WBSK2EVIAL
Innovative Grade US Origin Mouse C57BL6 Single Donor Whole Blood K2 EDTA Vial - Innovative Grade US Origin Mouse C57BL6 Single Donor Whole Blood from Innovative Research is collected per order from type C57BL6 Mice. The animals used for this material are of US Origin, ensuring the highest-quality material.