Learn More
Innovative Research Inc US Ms C57 Whl Bld Li Hep 25ml
Catalog No. 502744466
Encompass_Preferred
Innovative Grade US Origin Mouse C57BL6 Whole Blood Li Heparin 25ml - Innovative Grade US Origin Mouse C57BL6 Whole Blood from Innovative Research is collected per-order from type C57BL6 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used.
Catalog No. 50-274-4466 Supplier Innovative Research Inc Supplier No. IGMSC57WBLIH25ML
Innovative Grade US Origin Mouse C57BL6 Whole Blood Li Heparin 25ml - Innovative Grade US Origin Mouse C57BL6 Whole Blood from Innovative Research is collected per-order from type C57BL6 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used.