Learn More
Innovative Research Inc US Ms CD1 Comp Srm 1ml
Catalog No. 502744461
Encompass_Preferred
Innovative Grade US Origin Mouse CD1 Complement Serum 1ml - Innovative Grade US Origin Mouse CD1 Complement Serum from Innovative Research is manufactured using whole blood collections from type CD1 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.
Catalog No. 50-274-4461 Supplier Innovative Research Inc Supplier No. IGMSCD1CSER1ML
Innovative Grade US Origin Mouse CD1 Complement Serum 1ml - Innovative Grade US Origin Mouse CD1 Complement Serum from Innovative Research is manufactured using whole blood collections from type CD1 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.