Learn More
Innovative Research Inc US Ms CD1 Pla K3E 25ml
Catalog No. 502744456
Encompass_Preferred
Innovative Grade US Origin Mouse CD1 Plasma K3 EDTA 25ml - Innovative Grade US Origin Mouse CD1 Plasma from Innovative Research is manufactured using whole blood collections from type CD1 Mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.
Catalog No. 50-274-4456 Supplier Innovative Research Inc Supplier No. IGMSCD1PLAK3E25ML
Innovative Grade US Origin Mouse CD1 Plasma K3 EDTA 25ml - Innovative Grade US Origin Mouse CD1 Plasma from Innovative Research is manufactured using whole blood collections from type CD1 Mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.