Learn More
Innovative Research Inc US Ms CD1 Sngl Pla Li Hep Vl
Supplier:  Innovative Research Inc IGMSCD1PLASLIHVIAL
Encompass_Preferred
Innovative Grade US Origin Mouse CD1 Single Donor Plasma Li Heparin Vial - Innovative Grade US Origin Mouse CD1 Single Donor Plasma from Innovative Research is manufactured using whole blood collections from type CD1 Mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.
Catalog No. 50-274-4446