Learn More
Innovative Research Inc US Ms CD1 Whl Bld Li Hep 10ml
Catalog No. 502744442
Encompass_Preferred
Innovative Grade US Origin Mouse CD1 Whole Blood Li Heparin 10ml - Innovative Grade US Origin Mouse CD1 Whole Blood from Innovative Research is collected per-order from type CD1 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used.
Catalog No. 50-274-4442 Supplier Innovative Research Inc Supplier No. IGMSCD1WBLIH10ML
Innovative Grade US Origin Mouse CD1 Whole Blood Li Heparin 10ml - Innovative Grade US Origin Mouse CD1 Whole Blood from Innovative Research is collected per-order from type CD1 mice. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used.