Learn More
Innovative Research Inc US Pig Artery Carotid Frsh Sln
Catalog No. 502744435
Encompass_Preferred
Innovative Grade US Origin Porcine Artery Carotid Fresh in Saline - Innovative Grade US Origin Porcine Artery Carotid from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Porcine Carotid Artery measures about 4-5 inches in length and is processed from an animal approximately 6-9 months of age. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4435 Supplier Innovative Research Inc Supplier No. IGPCARCS
Innovative Grade US Origin Porcine Artery Carotid Fresh in Saline - Innovative Grade US Origin Porcine Artery Carotid from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Porcine Carotid Artery measures about 4-5 inches in length and is processed from an animal approximately 6-9 months of age. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).