Learn More
Innovative Research Inc US Pig Bile Fz 1L
Catalog No. 502744433
Encompass_Preferred
Innovative Grade US Origin Porcine Bile Frozen 1000ml - Innovative Grade US Origin Porcine Bile from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Porcine Bile is collected in the amount of approximately 30-50ml per animal. Large volumes provided as a pooled product. Custom volumes available upon request. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4433 Supplier Innovative Research Inc Supplier No. IGPCBILEZ1000ML
Innovative Grade US Origin Porcine Bile Frozen 1000ml - Innovative Grade US Origin Porcine Bile from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Porcine Bile is collected in the amount of approximately 30-50ml per animal. Large volumes provided as a pooled product. Custom volumes available upon request. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).