Learn More
Innovative Research Inc US Pig Buccal Mucosa Fz 1 Squ
Catalog No. 502744431
Encompass_Preferred
Innovative Grade US Origin Porcine Buccal Mucosa (Cheek) Frozen 1" Square - Innovative Grade US Origin Porcine Buccal Mucosa (Cheek) from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Size Options for this product include Standard 1" Square, Whole Inner Cheek (approximately 2"x3"), Whole Inner Cheek, Full Thickness, with Skin, Shaved (approximately 2"x3"), This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4431 Supplier Innovative Research Inc Supplier No. IGPCBUMZ1IN
Innovative Grade US Origin Porcine Buccal Mucosa (Cheek) Frozen 1" Square - Innovative Grade US Origin Porcine Buccal Mucosa (Cheek) from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Size Options for this product include Standard 1" Square, Whole Inner Cheek (approximately 2"x3"), Whole Inner Cheek, Full Thickness, with Skin, Shaved (approximately 2"x3"), This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).