Learn More
Innovative Research Inc US Pig Heart Heart Only Fz
Catalog No. 502744428
Encompass_Preferred
Innovative Grade US Origin Porcine Heart Heart Only Frozen - This Heart is collected on a custom basis from US Origin abattoir animals.Options IncludeHeart Only Heart Left Ventricle, Left Ventricle Only, With Aorta, With Abdominal Aorta, With Abdominal Aorta to Split, With Abdominal Aorta/Arteries, With Abdominal Aorta/Vena Cava, With Abdominal Aorta/Pulmonary Artery, With Abdominal Aorta/Pulmonary Artery, With Abdominal Aorta/Pulmonary Artery/Veins, With Descending Aorta, With Descending Aorta/Branches, With Descending Aorta/Vena Cava, With Pulmonary Artery. With Aortic Root, With Aorta/Veins/Arteries/Vena Cava, With Aorta/Inferior Vena Cava. This product is available freshor frozen.
Catalog No. 50-274-4428 Supplier Innovative Research Inc Supplier No. IGPCHTZT1
Innovative Grade US Origin Porcine Heart Heart Only Frozen - This Heart is collected on a custom basis from US Origin abattoir animals.Options IncludeHeart Only Heart Left Ventricle, Left Ventricle Only, With Aorta, With Abdominal Aorta, With Abdominal Aorta to Split, With Abdominal Aorta/Arteries, With Abdominal Aorta/Vena Cava, With Abdominal Aorta/Pulmonary Artery, With Abdominal Aorta/Pulmonary Artery, With Abdominal Aorta/Pulmonary Artery/Veins, With Descending Aorta, With Descending Aorta/Branches, With Descending Aorta/Vena Cava, With Pulmonary Artery. With Aortic Root, With Aorta/Veins/Arteries/Vena Cava, With Aorta/Inferior Vena Cava. This product is available freshor frozen.