Learn More
Innovative Research Inc US Pig Lymph Node Fresh
Catalog No. 502744423
Encompass_Preferred
Innovative Grade US Origin Porcine Lymph Node Fresh - Innovative Grade US Origin Porcine Lymph Node from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. The lymph node is generally collected from the neck and head. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4423 Supplier Innovative Research Inc Supplier No. IGPCLYNF
Innovative Grade US Origin Porcine Lymph Node Fresh - Innovative Grade US Origin Porcine Lymph Node from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. The lymph node is generally collected from the neck and head. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).