Learn More
Innovative Research Inc US Pig Vagina Fresh in Saline
Catalog No. 502744420
Encompass_Preferred
Innovative Grade US Origin Porcine Vagina Fresh in Saline - Innovative Grade US Origin Porcine Vagina from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Includes the vagina and cervix/vaginal cuff. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4420 Supplier Innovative Research Inc Supplier No. IGPCVAGS
Innovative Grade US Origin Porcine Vagina Fresh in Saline - Innovative Grade US Origin Porcine Vagina from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Includes the vagina and cervix/vaginal cuff. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).