Learn More
Innovative Research Inc US Rat Sprg Dwly Sngl Pla K2l
Catalog No. 502744396
Encompass_Preferred
Innovative Grade US Origin Rat Sprague Dawley Single Donor Plasma K2 EDTA Vial - Innovative Grade US Origin Rat Sprague Dawley Single Donor Plasma from Innovative Research is manufactured using whole blood collections from Sprague Dawley rats. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.
Catalog No. 50-274-4396 Supplier Innovative Research Inc Supplier No. IGRTSDPLASK2EVIAL
Innovative Grade US Origin Rat Sprague Dawley Single Donor Plasma K2 EDTA Vial - Innovative Grade US Origin Rat Sprague Dawley Single Donor Plasma from Innovative Research is manufactured using whole blood collections from Sprague Dawley rats. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.