Learn More
Innovative Research Inc US Rat Wstr Comp Srm 50ml
Catalog No. 502744389
Encompass_Preferred
Innovative Grade US Origin Rat Wistar Complement Serum 50ml - Innovative Grade US Origin Rat Wistar Complement Serum from Innovative Research is manufactured using whole blood collections from type Wistar rats. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.
Catalog No. 50-274-4389 Supplier Innovative Research Inc Supplier No. IGRTWICSER50ML
Innovative Grade US Origin Rat Wistar Complement Serum 50ml - Innovative Grade US Origin Rat Wistar Complement Serum from Innovative Research is manufactured using whole blood collections from type Wistar rats. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.