Learn More
Innovative Research Inc US Sh Bld Defibrin 1L
Catalog No. 502744378
Encompass_Preferred
Innovative Grade US Orgin Sheep Blood Defibrinated 1000ml - Innovative Grade US Origin Sheep Whole Blood Defibrinated from Innovative Research is collected per-order from sheep located in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The whole blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color.
Catalog No. 50-274-4378 Supplier Innovative Research Inc Supplier No. IGSHWBDF1000ML
Innovative Grade US Orgin Sheep Blood Defibrinated 1000ml - Innovative Grade US Origin Sheep Whole Blood Defibrinated from Innovative Research is collected per-order from sheep located in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The whole blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color.