Learn More
Innovative Research Inc US Sh Bld Defibrin 500ml
Catalog No. 502744377
Encompass_Preferred
Innovative Grade US Orgin Sheep Blood Defibrinated 500ml - Innovative Grade US Origin Sheep Whole Blood Defibrinated from Innovative Research is collected per-order from sheep located in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The whole blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color.
Catalog No. 50-274-4377 Supplier Innovative Research Inc Supplier No. IGSHWBDF500ML
Innovative Grade US Orgin Sheep Blood Defibrinated 500ml - Innovative Grade US Origin Sheep Whole Blood Defibrinated from Innovative Research is collected per-order from sheep located in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The whole blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color.