Learn More
Syndel USA Syndel Syncaine 10G
SDP
Supplier:  Syndel USA SYNCMGAUS03
Encompass_Preferred
Syndels Syncaine (also known as MS-222, TMS, tricaine methanesulfonate) is a fish anesthetic used for the temporary immobilization of fish, amphibians, and other aquatic cold-blooded animals (poikilotherms) around the world. Syncaine has long been recognized as a valuable tool for the proper handling of these animals during manual spawning (fish stripping), weighing, measuring, marking, surgical operations, transport, photography, and research. Syncaine is a fully approved drug in the USA, Canada and the United Kingdom.
Catalog No. NC2162657