Learn More
MSC Mechanix Wear SpeedKnit M-Pact Chemical Gloves, Sandy Nitrile-Coated, Medium, 1 ea
Catalog No. 5000516825
Encompass
These chemical resistant impact gloves feature an 18-gauge high-performance polyethylene and glass fiber blend knit designed for high dexterity and protection. They are engineered for demanding environments including oil and gas, marine, and fabrication applications.
  • Sandy nitrile-coated palms provide superior wet grip and high abrasion resistance
  • Dual-layered coating protects against harmful liquids and chemicals
  • Meets ANSI A4 cut resistance and ANSI level 4 abrasion standards
  • ANSI 138 level 2 impact protection for added safety
  • Gauntlet cuffs allow for easy on and off
  • Machine washable and dryable for convenience
Catalog No. 50-005-16825 Supplier MSC Supplier No. 06504815
These chemical resistant impact gloves feature an 18-gauge high-performance polyethylene and glass fiber blend knit designed for high dexterity and protection. They are engineered for demanding environments including oil and gas, marine, and fabrication applications.
  • Sandy nitrile-coated palms provide superior wet grip and high abrasion resistance
  • Dual-layered coating protects against harmful liquids and chemicals
  • Meets ANSI A4 cut resistance and ANSI level 4 abrasion standards
  • ANSI 138 level 2 impact protection for added safety
  • Gauntlet cuffs allow for easy on and off
  • Machine washable and dryable for convenience