Learn More
MSC Barcor Chamfer Gage, 3 to 4 in., 127 Degree, 1 ea.
Catalog No. 5000516846
Encompass
This O.D. chamfer gage is designed for precise measurement of chamfers and countersinks in the 3 in. to 4 in. range with a 127° angle. The direct reading instrument features a spring-loaded, conically ground, 3-blade gaging plunger for consistent accuracy. It is constructed from hardened tool steel and features a set-to number stamped on the back for calibration reference.
  • Capacity range of 3 in. to 4 in.
  • Hardened tool steel gaging plunger construction
  • Direct reading design with .001 in. resolution
  • Designed for 127° angular measurements
  • Spring-loaded 3-blade plunger mechanism
  • Made in the USA
Catalog No. 50-005-16846 Supplier MSC Supplier No. 06534960
This O.D. chamfer gage is designed for precise measurement of chamfers and countersinks in the 3 in. to 4 in. range with a 127° angle. The direct reading instrument features a spring-loaded, conically ground, 3-blade gaging plunger for consistent accuracy. It is constructed from hardened tool steel and features a set-to number stamped on the back for calibration reference.
  • Capacity range of 3 in. to 4 in.
  • Hardened tool steel gaging plunger construction
  • Direct reading design with .001 in. resolution
  • Designed for 127° angular measurements
  • Spring-loaded 3-blade plunger mechanism
  • Made in the USA