Learn More
MSC Vermont Gage Plus USA Class X Gage Pin, 0.1892 in., 2 in. Length, 1 ea.
Catalog No. 5000512329
Encompass
This Class X gage pin is a precision measuring instrument designed for the calibration of various measuring equipment. Manufactured from cold stabilized tool steel, this pin ensures durability and consistent performance in demanding metrology environments.
  • Features a diameter of 0.1892 in.
  • Hardness rated at 60/65 Rc for wear resistance
  • Finish of 4 microinch for high precision
  • NIST traceable for reliable measurement standards
  • Included certificate of accuracy
  • Roundness within 0.00002 in.
  • Tolerance of +0.00004 in./-0.00000 in.
  • Length of 2 in.
  • Made in the USA
Catalog No. 50-005-12329 Supplier MSC Supplier No. 93716157
This Class X gage pin is a precision measuring instrument designed for the calibration of various measuring equipment. Manufactured from cold stabilized tool steel, this pin ensures durability and consistent performance in demanding metrology environments.
  • Features a diameter of 0.1892 in.
  • Hardness rated at 60/65 Rc for wear resistance
  • Finish of 4 microinch for high precision
  • NIST traceable for reliable measurement standards
  • Included certificate of accuracy
  • Roundness within 0.00002 in.
  • Tolerance of +0.00004 in./-0.00000 in.
  • Length of 2 in.
  • Made in the USA