Learn More
USA LAB USA Lab Tri Clamp Tee, 3-Way, Fluoropolymer, 1.5 in. TC
Catalog No. NC3997337
Encompass
This PTFE/ETFE/PFA lined tri-clamp 3-way tee is ideal for high-purity and corrosive applications, featuring a seamless fluoropolymer bore and a stainless-steel body. It facilitates smooth directional splits and sampling with minimal hold-up, suitable for labs and industrial use. Available in 1.5 in. TC and 2 in. TC, media compatibility should be verified.
  • Fluoropolymer lined 304 stainless steel construction.
  • Tri-clamp sanitary design, CIP/SIP ready.
  • Seamless PTFE/ETFE/PFA bore minimizes adsorption/contamination.
  • Equal 3-way tee design for reduced turbulence and consistent process control.
  • Robust lining offers chemical resistance and extended service life.
  • Non-stick bore simplifies cleaning, reduces cross-contamination.
  • Typical operating range: ≈ −20°F to 400°F.
Catalog No. NC3997337 Supplier USA LAB Supplier No. 153WAYPTFE
This PTFE/ETFE/PFA lined tri-clamp 3-way tee is ideal for high-purity and corrosive applications, featuring a seamless fluoropolymer bore and a stainless-steel body. It facilitates smooth directional splits and sampling with minimal hold-up, suitable for labs and industrial use. Available in 1.5 in. TC and 2 in. TC, media compatibility should be verified.
  • Fluoropolymer lined 304 stainless steel construction.
  • Tri-clamp sanitary design, CIP/SIP ready.
  • Seamless PTFE/ETFE/PFA bore minimizes adsorption/contamination.
  • Equal 3-way tee design for reduced turbulence and consistent process control.
  • Robust lining offers chemical resistance and extended service life.
  • Non-stick bore simplifies cleaning, reduces cross-contamination.
  • Typical operating range: ≈ −20°F to 400°F.