Learn More
USA Scientific Inc 2.0 mL deep 96-well PP plate, sterile
Catalog No. NC9792727
Encompass
2.0 mL deep 96-well sterile PlateOne polypropylene plate, square wells, natural, individually wrapped, 20/pack
Catalog No. NC9792727 Supplier USA Scientific Inc Supplier No. 18962110
2.0 mL deep 96-well sterile PlateOne polypropylene plate, square wells, natural, individually wrapped, 20/pack