Learn More
Andwin Scientific USA SCIENTIFIC 5-50 5L 12 CHA
SDP
Catalog No. 50820512
Encompass_Preferred
USA Scientific 5-50 ul, 12 channel Adjustable Volume Multi-Channel Pipettor; Increments = 0.5ul; Accuracy = +/-1.00%; Precision = +/-0.50%
Catalog No. 50-820-512 Supplier Andwin Scientific Supplier No. 48120550
USA Scientific 5-50 ul, 12 channel Adjustable Volume Multi-Channel Pipettor; Increments = 0.5ul; Accuracy = +/-1.00%; Precision = +/-0.50%