Learn More
Ibidi USA µ-Slide 8 Well high Glass Bottom
SDP
Catalog No. NC2684473
Encompass
A chambered coverslip with 8 individual wells, high walls, and a #1.5H glass coverslip bottom, suitable for use in TIRF and single molecule applications. With a cell culture chamber for observation of the sample through a glass coverslip bottom using high resolution microscopy. Due to low thickness variability of the glass coverslip, you will get brilliant images of your cells. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: #1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, sterilizedPcs./Box: 90 (individually packed)
Catalog No. NC2684473 Supplier Ibidi USA Supplier No. 8080790
A chambered coverslip with 8 individual wells, high walls, and a #1.5H glass coverslip bottom, suitable for use in TIRF and single molecule applications. With a cell culture chamber for observation of the sample through a glass coverslip bottom using high resolution microscopy. Due to low thickness variability of the glass coverslip, you will get brilliant images of your cells. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: #1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, sterilizedPcs./Box: 90 (individually packed)