Learn More
MSC Vermont Gage Threaded Ring Gauge, 7/16-20 UNF, Class 2A, Go, 1 ea
Catalog No. 5000516795
Encompass
This threaded ring gauge is a precision inspection tool designed for verifying the pitch diameter and cumulative thread elements of externally threaded parts. Engineered for accuracy, this adjustable Go gauge ensures components meet the required specifications for Class 2A fit.
  • Constructed from durable tool steel for long-lasting wear resistance
  • Features a hardness range of 60-62 RC
  • Designed for Go (plus) inspection
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B1.2 standards
  • Compatible with 3T size holders
  • Uncoated finish
Catalog No. 50-005-16795 Supplier MSC Supplier No. 05423207
This threaded ring gauge is a precision inspection tool designed for verifying the pitch diameter and cumulative thread elements of externally threaded parts. Engineered for accuracy, this adjustable Go gauge ensures components meet the required specifications for Class 2A fit.
  • Constructed from durable tool steel for long-lasting wear resistance
  • Features a hardness range of 60-62 RC
  • Designed for Go (plus) inspection
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B1.2 standards
  • Compatible with 3T size holders
  • Uncoated finish