Learn More
Electron Microscopy Sciences USAF Test Chart R70 Positive Image
SDP
Catalog No. 501901572
Encompass_Preferred
USAF resolution charts are recognized the world over as a universal standard for testing the vertical and horizontal resolution of imaging systems. Each element on the chart comprises three vertical bars and three horizontal bars, and the detail on these slides is as fine as 0.78microns (644 line-pairs per mm). The resolution of the imaging system is normally specified as the Group and Element of the finest bars that can be clearly defined.
Type: Positive Image
Use: Groups 0 to 7
Catalog No. 50-190-1572 Supplier Electron Microscopy Sciences Supplier No. 6809801
USAF resolution charts are recognized the world over as a universal standard for testing the vertical and horizontal resolution of imaging systems. Each element on the chart comprises three vertical bars and three horizontal bars, and the detail on these slides is as fine as 0.78microns (644 line-pairs per mm). The resolution of the imaging system is normally specified as the Group and Element of the finest bars that can be clearly defined.
Type: Positive Image
Use: Groups 0 to 7