Learn More
MSC Mahr Data Connection Cable, USB, 2 m, 1 ea.
Catalog No. 5000514390
Encompass
This data collection cable and software package enables seamless connection between measuring instruments and a computer, facilitating efficient data transfer for recording and analysis. The kit is designed for use with Windows-based operating systems to streamline data integration into professional applications.
  • Connects measuring instruments to a PC, laptop, or tablet
  • Supports data transfer via virtual communication interface
  • Includes data connection cable, professional software, and driver
  • Compatible with multiple indicators
  • Features USB, simplex, and EX-interface hardware connections
Catalog No. 50-005-14390 Supplier MSC Supplier No. 88608328
This data collection cable and software package enables seamless connection between measuring instruments and a computer, facilitating efficient data transfer for recording and analysis. The kit is designed for use with Windows-based operating systems to streamline data integration into professional applications.
  • Connects measuring instruments to a PC, laptop, or tablet
  • Supports data transfer via virtual communication interface
  • Includes data connection cable, professional software, and driver
  • Compatible with multiple indicators
  • Features USB, simplex, and EX-interface hardware connections