Learn More
World Precision Instrument USB Drive 32GB, Programmed
SDP
Catalog No. 502521473
Encompass
This USB thumbdrive is loaded with the Python digital output package for use with the popular EVOM™ Manual TEER (transepithelial electrical resistance) meter. A blank replacement thumb drive is also available. This thumb drive hold 32 Gb of data and is suitable for use with the EVOM™ Manual for auto data logging of all your measurements. The retractable design of this SanDisk thumb drive protects the USB connector when the unit is not in use.
Catalog No. 50-252-1473 Supplier World Precision Instrument Supplier No. 300749
This USB thumbdrive is loaded with the Python digital output package for use with the popular EVOM™ Manual TEER (transepithelial electrical resistance) meter. A blank replacement thumb drive is also available. This thumb drive hold 32 Gb of data and is suitable for use with the EVOM™ Manual for auto data logging of all your measurements. The retractable design of this SanDisk thumb drive protects the USB connector when the unit is not in use.