Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ USB HAZ-LO Rechargeable Intrinsically Safe Headlamp, Spot/Flood

Catalog No. 19028340 Shop All Streamlight Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-028-340 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-028-340 Supplier Streamlight™ Supplier No. 61460
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This intrinsically safe headlamp is ideal for those in potentially hazardous work environments, with the added feature of USB rechargeability. With Class I, Div. 1 safety ratings, the dual beam headlamp has three output modes to cover a variety of tasks.

  • The USB HAZ-LO intrinsically safe headlamp is an ideal headlamp for those in potentially hazardous work environments, with the added feature of USB rechargeability.
  • With Class I, Div. 1 safety ratings, the dual beam headlamp has three output modes to cover a variety of tasks: Spot for a focused beam, Flood for tasks at arm's length and a Combo setting for even more light output
  • An extended runtime of 7 to 13 hours will last throughout your entire shift
  • Comes with both an elastic head strap and a rubber hard hat strap, as well as fasteners for strapless mounting to a hard hat
  • Class I, Div. 1 safety rated and USB rechargeable with charging indicator
  • Long runtimes of 7-13 hours that last throughout your work shift
  • Spot beam for a focused 89 foot beam at 190 lumens
  • Use flood beam mode for a soft 120 lumen light that won't tire your eyes
  • Spot/flood combo mode gives you 250 lumens for even more light
  • Multi-function push button switch easy to use, even with gloves
  • Easy to attach to hard hats via rubber strap or fasteners, which are included

Specifications

Voltage 3.7 V
Weight (English) 4.1 oz.
Weight (Metric) 0.116 kg
Battery Run Time 7 Hrs (High), 7.5 Hrs (Med), 13 Hrs (Low)
Battery Type Lithium-ion
Beam Distance 89 m
Candle Power 2000 cd
Certifications/Compliance Class I, II, Division 1, Groups A, B, C, D, E, F, G; Class III; CL 1, Zn 0, IIC; T-Code: T4 Meets applicable European Community Directives. CEC Compliant
Color Yellow
Description Rechargeable, Intrinsically Safe
Includes Rechargeable battery, 120 V AC charger, Elastic headstrap, Rubber hard hat strap, 3 M Dual Lock reclosable fasteners
Length (English) 1.75 in.
Lumen 250 lm (High), 190 lm (Med), 120 lm (Low)
Material Thermoplastic
Warranty 2 year warranty with proof of purchase
Product Type Rechargeable Headlamp
Show More Show Less

Class I, II, Division 1, Groups A, B, C, D, E, F, G; Class III; CL 1, Zn 0, IIC; T-Code: T4 Meets applicable European Community Directives. CEC Compliant

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.