Learn More
Julabo USA Inc Each, RS232 to USB type A cable
SDP
Catalog No. 507534276
Encompass_Preferred
Bath accessories, Julabo, USB Interface Adapter Cable (2.5 m) allows connection of JULABO instruments with RS232 interface to a PC with USB port. Included in delivery: USB Interface Cable RS232 9 pin USB Typ A. Adapter requires driver installation on PC.
Catalog No. 50-753-4276 Supplier Julabo USA Inc Supplier No. 8900110
Bath accessories, Julabo, USB Interface Adapter Cable (2.5 m) allows connection of JULABO instruments with RS232 interface to a PC with USB port. Included in delivery: USB Interface Cable RS232 9 pin USB Typ A. Adapter requires driver installation on PC.