Learn More
BINDER™ USB Interface Converter
SDP
Catalog No. 08111776 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Application):
APT-COM Software
Catalog No. For Use With (Application)
08-111-776 APT-COM Software
1 options
Catalog No. 08-111-776 Supplier BINDER™ Supplier No. 80120665
Add to Cart
Add to Cart
For Use With: Multi Management Software APT-COMTM
  • USB / RS422 - RS 422 cable set and RS 422 / USB interface converter for connection to 10-way plug distributor or for connection to a unit (USB-powered converter)
Specifications
Description Interface converter, USB / RS422 - RS 422 cable set and RS 422 / USB interface converter for connection to 10-way plug distributor or for connection to a unit (USB-powered converter)
For Use With (Equipment) Multi Management Software APT-COM™
Material Plastic
Type USB Powered Converter
For Use With (Application) APT-COM Software