Learn More
Cole-Parmer Oakton USB Data Cable for 450 and 2700 Meters
Supplier:  Cole-Parmer 3563053
Encompass_Preferred
Oakton USB Data Cable for 450 and 2700 Meters. Uses PL2303 UART chip
Catalog No. NC1249184