Learn More
Hach Company USB + Power Module for 2100Q/2100Q IS portable turbidimeters and DR1900 portable spectrophotometer
Catalog No. NC3862505 Shop All Hach Company Products
Encompass
The Hach USB + Power Module adds USB connectivity and mains power capability to compatible portable instruments. It enables XML data transfer to a PC, firmware updates, external printing, battery charging, and barcode-scanner-assisted data entry.
  • Adds USB connectivity for XML data transfer to a PC.
  • Enables operation from line power (100-240 Vac, 50/60 Hz).
  • Charges NiMH rechargeable batteries.
  • Supports firmware updates.
  • Allows printing to compatible external printers.
  • Facilitates barcode scanner input for quick sample and user IDs entry.
Catalog No. NC3862505 Supplier Hach Company Supplier No. LZV813.99.00002
The Hach USB + Power Module adds USB connectivity and mains power capability to compatible portable instruments. It enables XML data transfer to a PC, firmware updates, external printing, battery charging, and barcode-scanner-assisted data entry.
  • Adds USB connectivity for XML data transfer to a PC.
  • Enables operation from line power (100-240 Vac, 50/60 Hz).
  • Charges NiMH rechargeable batteries.
  • Supports firmware updates.
  • Allows printing to compatible external printers.
  • Facilitates barcode scanner input for quick sample and user IDs entry.