Learn More
Instech Moxa USB-to-serial conversion hub, 8 port. For use with Instech ABS1 or ABS2 automated blood samplers.
SDP
Catalog No. 5080976
Encompass
The Instech ABS1/USB08 is a specialized communication interface component used to connect Automated Blood Sampler™ (ABS™) units to a control computer. This MOXA-based USB-to-serial hub allows for the simultaneous management of multiple sampling units from a single workstation, ensuring reliable data transfer and synchronization during complex pharmacokinetic studies. It is a critical hardware link for scaling up research throughput in both rodent and large animal sampling labs.
Features
  • Application: USB communication interface for automated blood samplers
  • Compatibility: Works with ABS2™ systems and Instech control software
  • Capacity: Supports multi-unit configurations via 8-port conversion
  • Reliability: Industrial-grade hardware for stable data connection
  • Function: Synchronized control of up to 8 units per hub
  • Setup: Plug-and-play integration with ABS™ rack systems
Catalog No. 50-809-76 Supplier Instech Supplier No. ABS1/USB08
The Instech ABS1/USB08 is a specialized communication interface component used to connect Automated Blood Sampler™ (ABS™) units to a control computer. This MOXA-based USB-to-serial hub allows for the simultaneous management of multiple sampling units from a single workstation, ensuring reliable data transfer and synchronization during complex pharmacokinetic studies. It is a critical hardware link for scaling up research throughput in both rodent and large animal sampling labs.
Features
  • Application: USB communication interface for automated blood samplers
  • Compatibility: Works with ABS2™ systems and Instech control software
  • Capacity: Supports multi-unit configurations via 8-port conversion
  • Reliability: Industrial-grade hardware for stable data connection
  • Function: Synchronized control of up to 8 units per hub
  • Setup: Plug-and-play integration with ABS™ rack systems